Human Recombinant TrkA (from HEK293 Cells)
Supplier: ACRO Biosystems
Certificates
About this item
Human TrkA / NTRK1 (192-402) Protein, Mouse IgG2a Fc Tag, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 0.1 EU per μg by the LAL method.
- Gene ID:P04629-2
- Protein Synonyms:TRKA|Trk-A|MTC|NTRK1|TrkA|TRK
- Protein/Peptide Name:TrkA
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:49.1 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together