Human Recombinant NRG1 Beta 1 (from HEK293 Cells)
Human NRG1 Beta 1 Protein, Fc Tag, ACROBiosystems
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Gene ID:Q02297-6
- Protein Synonyms:Neuregulin1 beta 1|NDF|SMDF|ARIA|NRG1 Beta 1|NRG1|HRG1 beta 1|GGF|Neuregulin-1 beta 1|HRGA|HGL
- Protein/Peptide Name:NRG1 Beta 1
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:53.2 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT