Human Recombinant NKp46 (from HEK293 Cells)
Supplier: ACRO Biosystems
Certificates
About this item
Human NKp46 / NCR1 / CD335 Protein, His Tag, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Protein Synonyms:CD335|NCR1|hNKp46|NK-p46|LY94
- Protein/Peptide Name:NKp46
- Purity:>90% as determined by reduced SDS-PAGE.
- Molecular Weight:28.3 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together