Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant LIGHT (from HEK293 Cells)
Human Recombinant LIGHT (from HEK293 Cells)
Certificates
Human Recombinant LIGHT (from HEK293 Cells)

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Tag sequence:
    His Tag (active trimer) (MALS verified)
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Measured by its binding ability in a functional ELISA. Immobilized Human HVEM, Fc Tag (Cat. No. HVM-H5258) at 5 μg/mL (100 μL/well) can bind Human LIGHT, His Tag (active trimer) (MALS verified) (Cat. No. LIT-H5242) with a linear range of 16-63 ng/mL (QC tested). Measured by its binding ability in a functional ELISA. Immobilized Human DcR3, Fc Tag (Cat. No. TNB-H5255) at 5 μg/mL (100 μL/well) can bind Human LIGHT, His Tag (active trimer) (MALS verified) (Cat. No. LIT-H5242) with a linear range of 8-63 ng/mL (Routinely tested). Human LIGHT, His Tag (active trimer) (MALS verified) (Cat. No. LIT-H5242) induced cytotoxicity in HT-29 cells. The ED50 for this effect is 1.11-3.87 ng/mL (Routinely tested).
  • Gene ID:
    O43557-1
  • Protein Synonyms:
    HVEM-L|CD258|LTg|TR2|LIGHT|HVEML|CD258 antigen|TNFSF14
  • Protein/Peptide Name:
    Light
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    58.3 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications