Human Recombinant IL-5 (from HEK293 Cells)
Biotinylated Human IL-5 Protein, His,Avitag™, ACROBiosystems
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Species:Human
- Tag sequence:Precision Avi
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Gene ID:P05113-1
- Protein Synonyms:Interleukin-5|IL5|TRF|IL-5
- Protein/Peptide Name:IL-5
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:16.7 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS,pH7.3. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT