Human Recombinant IL-15 R alpha (from HEK293 cells), Biotin
Biotinylated Human IL-15 R alpha / CD215 Protein, Fc,Avitag™, ACROBiosystems
- Conjugation:Biotin
- Protein/Peptide Type:Recombinant
- Species:Human
- Tag sequence:Fc, Avitag™
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Human IL-15, Tag Free (Cat. No. IL5-H4117) at 2 μg/mL (100 μL/well) can bind Biotinylated Human IL-15 R alpha, Fc,Avitag (Cat. No. ILA-H82F4) with a linear range of 2-39 ng/mL (QC tested). Measured by its binding ability in a functional ELISA. Immobilized Mouse IL-15, Fc Tag (Cat. No. IL5-M5255) at 2 μg/mL (100 μL/well) can bind Biotinylated Human IL-15 R alpha, Fc,Avitag (Cat. No. ILA-H82F4) with a linear range of 10-156 ng/mL (Routinely tested).
- Gene ID:Q13261-1
- Protein Synonyms:CD215|IL-15RA|IL-15R-alpha|IL-15 R alpha|IL15RA
- Protein/Peptide Name:IL-15 R alpha
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:46.6 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in Tris with Glycine, Arginine and NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT