Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human Recombinant Her2 (from HEK293 cells)
 
Human Recombinant Her2 (from HEK293 cells)

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Species:
    Human
  • Tag sequence:
    Tag Free
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Measured by its binding ability in a functional ELISA. Immobilized Human Her2, Tag Free (Cat. No. HE2-H5212) at 1 μg/mL (100 μL/well)can bind Trastuzumab with a linear range of 0.2-2 ng/mL (QC tested). Loaded Hercecptin on AHC Biosensor, can bind Human Her2, Tag Free (Cat. No. HE2-H5212) with an affinity constant of 1.45 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    P04626-1
  • Protein Synonyms:
    NEU|MLN19|TKR1|ERBB2|CD340|HER2|HER-2/neu|NGL
  • Protein/Peptide Name:
    Her2
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    70.1 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT