Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Human Recombinant Complement Component C2 (from HEK293 Cells)

Marketsource
Human Recombinant Complement Component C2 (from HEK293 Cells)
Marketsource
 
Human Recombinant Complement Component C2 (from HEK293 Cells)

 

 

Human Complement Component C2 Protein, His Tag (MALS verified) (active enzyme), ACROBiosystems

 

 

 

  • Catalog No:
  • 103814-966
  • 103814-964
  • Protein/Peptide Name:
  • Complement Component C2
  • Complement Component C2
  • Protein Synonyms:
  • Complement Component C2|Complement C2|C2
  • Complement Component C2|Complement C2|C2
  • Protein/Peptide Type:
  • Recombinant
  • Recombinant
  • Species:
  • Human
  • Human
  • Source:
  • HEK293 cells
  • HEK293 cells
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Gene ID:
  • P06681-1
  • P06681-1
  • Endotoxin Level:
  • Low
  • Low
  • Endotoxin Content:
  • Less than 1.0 EU per μg by the LAL method.
  • Less than 1.0 EU per μg by the LAL method.
  • Purity:
  • >90% as determined by SDS-PAGE.
  • >90% as determined by SDS-PAGE.
  • Formulation:
  • Lyophilized from 0.22 μm filtered solution in 20 mM Tris, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Lyophilized from 0.22 μm filtered solution in 20 mM Tris, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Molecular Weight:
  • 83.0 kDa
  • 83.0 kDa
  • Storage Conditions:
  • –20 °C
  • –20 °C
  • Shipping Temperature:
  • RT
  • RT

 

Product Family Options

 

  • Size
     
     
  • 50 µg
       
       
    Each (50µG)
    Each (50µG) - $432.00
    $432.00
    Each (50µG)
       $432.00
     

     : CO2H52H750UG
     
     
    Marketsource
     
    Specifications
    Cat. No.
    103814-966
    Protein/Peptide Name
    Complement Component C2
    Protein Synonyms
    Complement Component C2|Complement C2|C2
    Protein/Peptide Type
    Recombinant
    Species
    Human
    Source
    HEK293 cells
    Conjugation
    Unconjugated
    Gene ID
    P06681-1
    Endotoxin Level
    Low
    Endotoxin Content
    Less than 1.0 EU per μg by the LAL method.
    Purity
    >90% as determined by SDS-PAGE.
    Formulation
    Lyophilized from 0.22 μm filtered solution in 20 mM Tris, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
    Molecular Weight
    83.0 kDa
    Storage Conditions
    –20 °C
    Shipping Temperature
    RT
    Marketsource
  • 1 mg
       
       
    Each (1mg)
    Each (1mg) - $3,534.00
    $3,534.00
    Each (1mg)
       $3,534.00
     

     : CO2H52H71MG
     
     
    Marketsource
     
    Specifications
    Cat. No.
    103814-964
    Protein/Peptide Name
    Complement Component C2
    Protein Synonyms
    Complement Component C2|Complement C2|C2
    Protein/Peptide Type
    Recombinant
    Species
    Human
    Source
    HEK293 cells
    Conjugation
    Unconjugated
    Gene ID
    P06681-1
    Endotoxin Level
    Low
    Endotoxin Content
    Less than 1.0 EU per μg by the LAL method.
    Purity
    >90% as determined by SDS-PAGE.
    Formulation
    Lyophilized from 0.22 μm filtered solution in 20 mM Tris, 150 mM NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
    Molecular Weight
    83.0 kDa
    Storage Conditions
    –20 °C
    Shipping Temperature
    RT
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.