Human Recombinant CD7 (from HEK293 Cells), FITC (Fluorescein Isothiocyanate)
Supplier: ACROBIOSYSTEMS INC MS
Certificates
About this item
FITC-Labeled Human CD7 Protein, Llama IgG2b Fc Tag, ACROBiosystems
Specifications
- Protein/Peptide Type:Recombinant
- Species:Human
- Storage Conditions:–20 ℃, aviod light
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Gene ID:P09564-1
- Protein Synonyms:LEU-9|TP41|CD7|Tp40|GP40
- Protein/Peptide Name:CD7
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:44.3 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT