Human Recombinant CD59 (from HEK293 Cells)
Supplier: ACROBIOSYSTEMS INC MS
Certificates
About this item
Human CD59 Protein, His Tag, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Gene ID:P13987-1
- Protein Synonyms:Protectin|FLJ38134|MIRL|MEM43|1F5|MACIF|MIN3|EJ16|MSK21|EL32|MGC2354|HRF20|p18-20|MIC11|MIN1|CD59|16.3A5|MIN2|MAC-IP|G344|EJ30|FLJ92039
- Protein/Peptide Name:CD59
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:10.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT