Hepacivirus C Recombinant E2 (from HEK293 cells)
Supplier: ACROBIOSYSTEMS INC MS
Certificates
About this item
Hepacivirus C E2 Protein, His Tag, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Hepacivirus C
- Tag sequence:His Tag
- Storage Conditions:–20 °C
- Gene ID:Q0N1H1-1
- Protein/Peptide Name:E2
- Molecular Weight:22.6 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT