Cynomolgus Monkey Recombinant Siglec-10 (from HEK293 cells)
Supplier: ACROBIOSYSTEMS INC MS
Certificates
About this item
Cynomolgus Siglec-10 Protein, His Tag, ACROBiosystems
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Gene ID:A0A2K5WBX8-1
- Protein Synonyms:Siglec10|PRO940|SIGLEC10|SLG2|MGC126774
- Protein/Peptide Name:Siglec-10
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:60.2 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in 25 mM MES, 150 mM NaCl, pH5.5. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT