Order Entry
Puerto Rico
ContactUsLinkComponent
SmartWall® Flat Grid Shelf, Metro™
SmartWall® Flat Grid Shelf, Metro™
Catalog # 76463-114
Supplier:  Metropolitan Wire
undefined
SmartWall® Flat Grid Shelf, Metro™
Catalog # 76463-114
Supplier:  Metropolitan Wire
Supplier Number:  FGS1224K4

Specifications

  • Finish:
    Metroseal epoxy
  • Width:
    12¹¹⁄₁₆"
  • Color:
    Gray
  • Description:
    Flat grid shelf
  • Height:
    4"
  • Length:
    24"
  • Material:
    Steel
  • Cat. No.:
    76463-114

Specifications

About this item

Keep small supplies and tools within reach.

  • Provides easy access to small items
  • Attaches directly to SmartWall® grids without the use of tools
  • Wire design makes it suitable for drying

Metro® SmartWall® Flat Grid Shelves are a great way to keep small items off of your workspace.

The Microban® antimicrobial product protection works to shield microorganisms like bacteria, mold and mildew from damaging the shelf. Metroseal epoxy coating also ensures that the shelf will not corrode in wet environments.