Diversey™ Oxivir® TB Disinfectant Wipes, Essendant
Supplier: Janitorial Supplies
Certificates
About this item
Ready-to-use disinfectant cleaner wipes are based on proprietary hydrogen peroxide (AHP®) technology to deliver a fast, effective, general-purpose cleaning performance.
- Disinfects in 60 seconds
- Kills MRSA and Norovirus
- Chemical compound: Benzyl Alcohol
- No VOCs and no fragrance added
- Non-irritating to skin and eyes
Oxivir® TB Disinfectant Wipes work as a virucide, bactericide, tuberculocide, fungicide, and non-food contact sanitizer. These wipes meet bloodborne pathogen standards for decontaminating blood and body fluids. Use for general-purpose applications on hard, non-porous surfaces.