Anti-DNP HEK293 Polyclonal Antibody (N297A)
Supplier: ACRO Biosystems
Certificates
About this item
Type: Primary
Antigen: DNP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: HEK293
Isotype: Human IgG1 (N297A)/kappa
Reactivity:
Specifications
- Catalog No:
- 103815-536
- 103815-538
- Antigen Symbol:
- DNP
- DNP
- Antigen Name:
- DNP
- DNP
- Antigen Synonyms:
- DNP,Dinitrophenyl
- DNP,Dinitrophenyl
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Host:
- HEK293
- HEK293
- Form:
- Powder
- Powder
- Purification:
- Affinity purified
- Affinity purified
- Shipping Temperature:
- RT
- RT
Specifications
Frequently Bought Together