Diversey™ Virex® II 256 One-Step Disinfectant Cleaner Deodorant, Mint, Essendant
Supplier: Janitorial Supplies
Certificates
About this item
A one-step, quaternary-based disinfectant cleaner and deodorant for use in facilities where cleaning and prevention of cross-contamination are critical. Neutral, no-rinse formula will not dull, pit or soften floor finishes. Bactericidal, virucidal and fungicidal. Meets hospital standards for decontaminating blood and body fluids.
- For use in facilities where cleaning and prevention of cross-contamination are critical
- Neutral, no-rinse formula will not dull, pit or soften floor finishes
- Bactericidal, virucidal and fungicidal
- Meets hospital grade standards for decontaminating blood and body fluids
- Mint scent
Application: Disinfectant/Cleaner.
Applicable Material: Ceramics; Enamel; Fiberglass; Finished/Sealed Wood; Laminates; Metal; Plastic; Sealed/Engineered Stone; Vinyl.
Dirt Types: Bacteria; Dust; Fungus; Germs; Grime; Mildew; Mold; Organic Matter; Viruses; Water-Based Soil.