Order Entry
Puerto Rico
ContactUsLinkComponent
Oxivir® 1 RTU Disinfectant Cleaner, Essendant
Oxivir® 1 RTU Disinfectant Cleaner, Essendant
Catalog # 76461-966
undefined
Oxivir® 1 RTU Disinfectant Cleaner, Essendant
Catalog # 76461-966
Supplier Number:  DVO100850916

Specifications

  • Description:
    Oxivir® 1 RTU disinfectant cleaner
  • Size:
    12 × 32 oz.
  • Cat. No.:
    76461-966
  • Packaging:
    Spray bottle

Specifications

About this item

Next generation ready-to-use disinfectant cleaner formulated with patented hydrogen peroxide technology (AHP®) to deliver fast, effective cleaning performance. Contains no phosphates, quat or phenols. Cleans, disinfects and deodorizes in one labor-saving step. It is a one minute bactericidal, fungicidal, tuberculocidal and 10 sec non-food contact sanitizer. Disinfects viruses and is a soft surface sanitizer, both in 30 seconds. Kills MRSA, Norovirus, RSV and Adenovirus. Meets bloodborne pathogen standards for decontaminating surfaces soiled with blood and body fluids. Destroys allergens and kills germs, while cleaning hard, nonporous surfaces; removes allergens like pet dander, cockroach matter, dust mite debris, and pollen particles.

  • Disinfectant cleaner formulated with patented hydrogen peroxide technology (AHP®) to deliver effective cleaning
  • Contains no phosphates, quat or phenol
  • Cleans, disinfects and deodorizes in one step
  • Broad spectrum, multi-surface bactericidal, fungicidal, tuberculocidal and 10 second non-food contact sanitizer
  • Disinfects viruses and is a soft surface sanitizer, both in 30 seconds
  • Kills MRSA, Norovirus, RSV and Adenovirus; meets bloodborne pathogen standards for decontaminating surfaces soiled with blood and body fluids
  • Harnesses the cleaning power of hydrogen peroxide; specially designed to penetrate the toughest soils

One-step cleaner, disinfectant, and deodorizer on hard, nonporous surfaces with no rinsing required. Harnesses the cleaning power of hydrogen peroxide; specially designed to penetrate the toughest soils. Suitable for everyday use; compatible with virtually all surfaces (avoid contact with brass and copper).

Application: Disinfectant/Cleaner.

Applicable Material: Hard, Non-Porous Surfaces.

Chemical Compound: Benzyl Alcohol.

Dirt Types: Bacteria; Fungus; Viruses.