Order Entry
Puerto Rico
ContactUsLinkComponent
 
Diversey™ Detergent/Disinfectant, Lemon Scent
Catalog # 76462-010
undefined
Diversey™ Detergent/Disinfectant, Lemon Scent
Catalog # 76462-010
Supplier Number:  DVO5412135

Specifications

  • Description:
    Detergent/Disinfectant, Lemon Scent
  • Size:
    2×90 packs (5 oz).
  • Cat. No.:
    76462-010
  • Packaging:
    Packet
  • Form:
    Powder

Specifications

About this item

Premeasured water-soluble packet takes the guesswork out of cleaning. Cleans, deodorizes and disinfects hard surfaces. Effective against mildew, bacteria and viruses including HIV-1 (AIDS virus). Idea for use in foodservice, nursing homes and other settings where housekeeping and disinfection are important.

  • Premeasured and water-soluble, so no guesswork, waste or messy clean-up
  • Cleans, disinfects and deodorizes hard surfaces such as walls, floors, sink tops, tables and chairs
  • Effective against mildew, bacteria and viruses including HIV-1 (AIDS virus)
  • Ideal for use in foodservice, nursing homes
  • Lemon scent

Application: Disinfectant/Cleaner.

Applicable Material: Brick; Cement; Ceramics; Concrete; Enamel; Fiberglass; Finished/Sealed Wood; Glass; Laminates; Metal; Plaster; Plastic; Vinyl.

Dirt Types: Bacteria; Dust; Fungus; Germs; Grime; Mildew; Viruses; Water-Based Soil.

Ordering information: 90 packets per tub, two tubs per carton.