Order Entry
Puerto Rico
ContactUsLinkComponent
 
Diversey™ Crew® Restroom Floor and Surface SC Non-Acid Disinfectant Cleaner, Essendant
  76461-986
undefined
Diversey™ Crew® Restroom Floor and Surface SC Non-Acid Disinfectant Cleaner, Essendant
  76461-986
 :  DVO101102189

 

  • Description:
    Crew® Restroom Floor & Surface SC Non-Acid Disinfectant Cleaner
  • Size:
    2× 1.4 L
  • Cat. No.:
    76461-986
  • Packaging:
    Bottle, SmartDose®
  • Form:
    Liquid

 

 

Non-acid restroom disinfectant cleaner. Cleans, disinfects and deodorizes in one easy step. Recommended for hard, nonporous, washable surfaces in and out of the restroom. Utilizes Crew® fresh scent to keep consistent with most Crew® restroom products.

  • No rinsing required for non-food contact surfaces.
  • Provides broad spectrum efficacy against common bathroom microoragnisms.
  • Kills HIV-1, Staphylococcus aureus, Salmonella choleraesuis, E. coli, MRSA, VRE and other microorganisms.
  • Deodorizes as it disinfects
  • Apply by mop and bucket, trigger sprayer or sponge

Application: Disinfectant/Cleaner.

Applicable Material: Nonporous Bathroom Surfaces.

Chemical Compound: Alcohol Ethoxylates.

Dirt Types: HIV-1; Staphylococcus aureus; Salmonella choleraesuis; E. coli; MRSA; VRE.