ContactUsLinkComponent
Avantor® ACE®, Replacement Ferrules
Avantor® ACE®, Replacement Ferrules
Catalog # 76408-450
Supplier:  Avantor
   Low Manufacturing Impact
Avantor® ACE®, Replacement Ferrules
Catalog # 76408-450
Supplier:  Avantor
Supplier Number:  F0001
   Low Manufacturing Impact
Specifications
  • Description:
    Replacement PEEK ferrules for UHPLC guard cartridge holder for 2,1 to 4,6 mm ID columns
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental Certification:
    ISO 14001:2015 (530840 UM15)
  • Cat. No.:
    76408-450