About this item
A ready-to-use, quaternary-based, hospital-grade disinfectant that provides excellent cleaning and deodorizing in one step. Disinfects in three minutes. Bactericide, tuberculocide, virucide and fungicide. Kills Norovirus, VRE and MRSA. For use on hard, inanimate, non-porous surfaces such as floors, walls, porcelain and plastic. This product has demonstrated effectiveness against SARS-CoV-2 (the Novel Coronavirus that causes COVID-19) on hard non-porous surfaces in just 1 minute.
- One-step quaternary disinfectant cleaner and deodorant to clean and disinfect hard surfaces in schools, offices, hotels, hospitals, nursing homes, and other facilities
- Has demonstrated effectiveness against SARS-CoV-2 (the Novel Coronavirus that causes COVID-19) on hard non-porous surfaces in just 1 minute
- Provides broad-spectrum kill of microorganisms
- Meets OSHA's Bloodborne pathogen standards for HBV and HIV
- Kills the organisms that cause odors and works as an odor counteractant to further eliminate odors not associated with bacteria
- Citrus scent, colorless
- Can be applied with a mop, sponge, or cloth
Provides broad-spectrum kill of microorganisms including HBV, HIV-1, VRE, MRSA, GRSA, MRSE, VISA, PRSP, Herpes Simplex Types 1&2, Influenza Type A2, Adenovirus, Rotavirus, and many more.
Application: Hard surfaces; Nonporous surfaces.
Applicable Material: Floors; Plastic; Porcelain; Walls
Dirt Types: Bacteria; Fungus; MRSA; Tuberculosis; VRE; Virus.
Chemical Compound: Diethylene Glycol Butyl Ether.