About this item
The NEBNext® RNA depletion core reagent set provides a customized approach to deplete unwanted RNA from any organism, using probe sequences designed with the user-friendly NEBNext® custom RNA depletion design tool.
- Compatible with a broad range of input amounts: 10 ng to 1 µg
- Suitable for low-quality or high-quality RNA
- Fast workflow: Two hours, with less than ten minutes hands-on time
In RNA-seq, highly expressed transcripts with minimal biological interest, such as ribosomal RNA (rRNA) can dominate readouts and mask detection of more informative low-abundance transcripts. This challenge is amplified when working with sample types for which pre-designed RNA depletion kits are not available.
The efficient RNase-H-based workflow, and close tiling of probes designed using the online tool, enables effective depletion from both low- and high-quality RNA, with a broad range of input amounts.
The concentration is not applicable for NEBNext® RNA depletion core reagent set.
Specifications
- Method / Format:Enzymatic clean-up
- Prep size:Scalable
- Refine target molecule:Total RNA
- Sample type:RNA preps
- Target molecule:RNA
- Description:NEBNext® RNA depletion core reagent set