Order Entry
Puerto Rico
ContactUsLinkComponent
Lovibond® UV/Visible Spectrophotometers, XD 7000 (Vis) and XD 7500 (UV-Vis), Lovibond Tintometer
Lovibond® UV/Visible Spectrophotometers, XD 7000 (Vis) and XD 7500 (UV-Vis), Lovibond Tintometer
Supplier:  Lovibond
Certificates
undefined
Lovibond® UV/Visible Spectrophotometers, XD 7000 (Vis) and XD 7500 (UV-Vis), Lovibond Tintometer
Supplier:  Lovibond

Specifications

  • Catalog No:
  • 76452-166
  • 76452-164
  • Model:
  • XD 7500, Grid monochromator
  • XD 7000, Grid monochromator
  • Measuring Modes:
  • Concentration, single and multi-wavelength measurement of absorbance and % transmission, kinetics, spectra
  • Concentration, single and multi-wavelength measurement of absorbance and % transmission, kinetics, spectra
  • Display:
  • 7" high contrast colour graphic-display
  • 7" high contrast colour graphic-display
  • Optical design:
  • Grid monochromator with reference beam and beam splitter after exit slit
  • Grid monochromator with reference beam and beam splitter after exit slit
  • Photometric Accuracy:
  • 0.003 Abs below 0.6 Abs; 0.5% from 0.6 to 2.0 Abs
  • 0.003 Abs below 0.6 Abs; 0.5% from 0.6 to 2.0 Abs
  • Photometric Linearity:
  • <1% up to 2.0 Abs between 340 to 900 nm
  • <1% up to 2.0 Abs between 340 to 900 nm
  • Photometric Range:
  • −3.3 - +3.3 Abs
  • −3.3 - +3.3 Abs
  • Photometric Reproducibility:
  • 0.003 Abs below 0.6 Abs; 0.5% from 0.6 to 2.0 Abs
  • 0.003 Abs below 0.6 Abs; 0.5% from 0.6 to 2.0 Abs
  • Photometric Resolution:
  • Absorption: 0.001 ; Transmission: 0.1%
  • Absorption: 0.001 ; Transmission: 0.1%
  • Wavelength Accuracy:
  • ±1 nm on all holmium peaks
  • ±1 nm on all holmium peaks
  • Wavelength Reproducibility:
  • better than 0.5 nm
  • better than 0.5 nm
  • Datalog - Capacity (Sets):
  • 5000 approx.
  • 5000 approx.
  • Electrical:
  • 100 - 240 V, 50/60 Hz
  • 100 - 240 V, 50/60 Hz
  • Languages:
  • German, English, French, Spanish, Italian, Portuguese, Polish, Indonesian, Russian, Chinese, Japanese, Dutch, Swedish, Norwegian, Czech, Romanian, Macedonian, Slovenian, Hungarian, Turkish, Korean, Vietnamese, Thai, Serbian, Malaysian, Danish, Bulgarian
  • German, English, French, Spanish, Italian, Portuguese, Polish, Indonesian, Russian, Chinese, Japanese, Dutch, Swedish, Norwegian, Czech, Romanian, Macedonian, Slovenian, Hungarian, Turkish, Korean, Vietnamese, Thai, Serbian, Malaysian, Danish, Bulgarian
  • Power input:
  • Buffer batteries (4×AA), power supply unit with cable
  • Buffer batteries (4×AA), power supply unit with cable
  • Spectral bandwidth:
  • 4 nm
  • 4 nm
  • Stray Light:
  • <0.05% transmission at 340 and 408 nm
  • <0.1% transmission at 340 and 408 nm
  • Dimensions:
  • 422×195×323 mm
  • 422×195×323 mm
  • Weight:
  • 4.5 kg
  • 4.5 kg

Specifications

Frequently Bought Together