ContactUsLinkComponent
EDX/XEDS Calibration TEM Window Grid, Electron Microscopy Sciences
EDX/XEDS Calibration TEM Window Grid, Electron Microscopy Sciences
Catalog # 76439-544
EDX/XEDS Calibration TEM Window Grid, Electron Microscopy Sciences
Catalog # 76439-544
Supplier Number:  76042-01
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    EDX-XEDS TEM window grid
  • Cat. No.:
    76439-544
Specifications
About this item
Suspended germanium provides a unique calibration standard for x-ray energy dispersive spectroscopy. Since Ge is not typically found in TEM columns, the calibration samples provide a material that cannot be mistaken for instrument components and their signal peaks. The regime in which system peaks normally occur (2 to 9 keV and 11 to 20 keV) is devoid of peaks from the Ge.
  • 20 nm thick germanium (Ge) coating on microporous 20 nm thick, low-stress silicon nitride (SiN)
  • 2 µm pores on 1:1 pitch grid pattern
  • 100 µm thick frame, fits 3 mm sample holders
  • (1) 500×500 µm window
The Ge is suspended across 2 µm pores that are patterned on a grid of 20 nm thick silicon nitride.
The single 500×500 µm window is compatible with high tilt angle tomography, since at 70° of tilt, the thin and beveled 100 µm silicon frame allows you to use a ~50×50 µm region within the center of the window from any rotational orientation.
These EDX calibration standards were developed in partnership with Dr. Nestor J. Zaluzec from the Electron Microscopy Center and the Center for Nanoscale Materials at Argonne National Laboratory.
Applications in detector energy axis and energy resolution calibration, detector window transmission evaluation, detector solid angle measurements, electron optical instrument system peak measurements, and specimen.