Order Entry
Puerto Rico
ContactUsLinkComponent
Tritirachium album Limber Proteinase K
Tritirachium album Limber Proteinase K
Catalog # RLMB-112-0100
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Tritirachium album Limber Proteinase K
Catalog # RLMB-112-0100
Supplier Number:  MB-112-0100
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Specifications

  • Species:
    Tritirachium album limber
  • Size:
    100 mg
  • Enzyme Name:
    Proteinase K
  • Cat. No.:
    RLMB-112-0100
  • Supplier no.:
    MB-112-0100

Specifications

About this item

All enzymes and related reagents are configured as an integrated system and have been thoroughly tested.

Application: Structural analysis in molecular cloning, DNA sequencing, Nucleic acid research, and protein/peptide sequencing.