Order Entry
Puerto Rico
ContactUsLinkComponent
 
Flammable
Irritant
Environment
Sodium dodecyl sulfate (SDS), PlusOne™
Catalog # 95017-116
Supplier:  Cytiva
Product available on GSA Advantage®
CAS Number:  
undefined
Flammable
Irritant
Environment
Sodium dodecyl sulfate (SDS), PlusOne™
Catalog # 95017-116
Supplier:  Cytiva
Supplier Number:  17-1313-01
Product available on GSA Advantage®
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle for solids
  • Size:
    100 g
  • Cat. No.:
    95017-116
  • Supplier no.:
    17-1313-01

Specifications

About this item

 :
 :288.38 g/mol
Melting Pt:204…207 °C
Storage Temperature:Ambient
UN:8027
ADR:4.1,III
 :GHS02
 :GHS07
 :GHS09
CAS Number:151-21-3
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175