Order Entry
Puerto Rico
ContactUsLinkComponent
EagI-HF®, New England Biolabs
EagI-HF®, New England Biolabs
Catalog # 101641-190
Small Business Enterprise
CAS Number:  
undefined
EagI-HF®, New England Biolabs
Catalog # 101641-190
Supplier Number:  R3505M
Small Business Enterprise
CAS Number:  

Specifications

  • Concentration:
    100000 U/ml
  • Description:
    EagI-HF®
  • Size:
    2500 units
  • Enzyme Name:
    EagI-HF
  • Cat. No.:
    101641-190
  • Supplier no.:
    R3505M

Specifications

About this item

An E.coli strain that carries the cloned and modified EagI gene from Enterobacter agglomerans (R. Morgan).

High Fidelity (HF®) Restriction Enzymes have 100% activity in CutSmart® Buffer; single-buffer simplicity means more straightforward and streamlined sample processing. HF enzymes also exhibit dramatically reduced star activity. HF enzymes are all Time-Saver™ qualified and can therefore cut substrate DNA in 5-15 with the flexibility to digest overnight without degradation to DNA. Engineered with performance in mind, HF restriction enzymes are fully active under a broader range of conditions, minimizing off-target products, while offering flexibility in experimental design.