ContactUsLinkComponent
Monarch® RNA Lysis Buffer, New England Biolabs
Monarch® RNA Lysis Buffer, New England Biolabs
Catalog # 103529-152
CAS Number:  
Monarch® RNA Lysis Buffer, New England Biolabs
Catalog # 103529-152
Supplier Number:  T2012L
CAS Number:  
Specifications
  • Description:
    Monarch® RNA lysis buffer
  • Size:
    100 ml
  • Cat. No.:
    103529-152
Specifications
About this item
Need more RNA Lysis Buffer? No problem. Monarch® Kit components are all available for purchase separately.
  • Can be used to lyse cells and release cellular components, including RNA
The Monarch® RNA Lysis Buffer is a component of the Monarch® Total RNA Miniprep Kit.
Delivery: Monarch® RNA Lysis Buffer is supplied in two bottles each containing 50 ml.