ContactUsLinkComponent
Monarch® gDNA Binding Buffer, New England Biolabs
Monarch® gDNA Binding Buffer, New England Biolabs
Catalog # 76339-780
CAS Number:  
Monarch® gDNA Binding Buffer, New England Biolabs
Catalog # 76339-780
Supplier Number:  T3014L
CAS Number:  
Specifications
  • Description:
    Monarch® gDNA binding buffer
  • Size:
    65 ml
  • Cat. No.:
    76339-780
Specifications
About this item
The Monarch® gDNA Binding Buffer a component of the Monarch® Genomic DNA Purification Kit (NEB #T3010), which can be used to purify up to 30 µg genomic DNA from a variety of sample type.
The Monarch® gDNA Binding Buffer is a chaotropic salt-based binding buffer with low alcohol content, designed to optimize binding of gDNA to the spin column. The binding of gDNA takes place at low speed, maximizing binding and efficiency.