ContactUsLinkComponent
Monarch® gDNA Cell Lysis Buffer, New England Biolabs
Monarch® gDNA Cell Lysis Buffer, New England Biolabs
  76339-776
 :  
Monarch® gDNA Cell Lysis Buffer, New England Biolabs
  76339-776
 :  T3012L
 :  
  • Description:
    Monarch® gDNA cell lysis buffer
  • Size:
    20 ml
  • Cat. No.:
    76339-776
The Monarch® gDNA Cell Lysis Buffer is a component of the Monarch® Genomic DNA Purification Kit (NEB #T3010), which can be used to purify up to 30 µg genomic DNA from a variety of sample types.
  • Provides mild lysis conditions that help reduce the viscosity that is common in cell samples
The Monarch® gDNA Cell Lysis Buffer is one of three lysis buffers available for purchase separately for additional convenience and to minimize waste.
Optimized for speed and efficiency, this buffer requires only a 5 minute lysis incubation time with Monarch® Proteinase K and Monarch® RNase A to obtain maximal yields of highly pure and intact gDNA with a typical peak size >60 kb.