Pre and Post Vacuum Autoclave, 3870EL-PV-G-WS
77041-316
- Model:5075EL-PV-G-WS
- Height:17"
- Plug Type:NEMA L15-30
- Cat. No.:77041-316
- Ext. dimensions:34.3×46.3×62.1"
- Electrical:220 - 240 V, 60 Hz, 3 phase
- Chamber volume:160 L
- Chamber dimensions:19.7×29.5"
The 3870EL-PV-G-WS can be operated as a stationary sterilizer connected to the builidings utilities for distilled water, tap water, drain and electricity.
- Pre and Post Vacuum for faster more efficient air removal (tubing, glassware, pouches, wrapped tools and garments)
- Built-in steam generator and vacuum pump for complete drying
- Water saving system
- Free standing with built-in swivel wheels for mobility
- Optional Biohazard filter (BH) prevents contaminated air from exhausting into the lab
The units come as standard with 'Plug and Play' option to operate with an electric power connection. In this mode there is no need to connect the autoclave to the building utilites for distilled water, tap water and drain.
: Model 3870EL-PV-G-WS has a load capacity of 23×250 ml, 17×500 ml, 10×1000 ml or 5×2000 ml Erlenmeyer flasks or 36×250 ml, 23x500 ml, 18×1000 ml or 9×2000 ml Schott Duran flasks.
Model 5075EL-PV-G-WS has a load capacity of 1×50 and 1×18 250 ml, 1×18, and 1×18 500 ml, 15×1000 ml, 7×2000 ml, 4×3000 ml or 3×5000 ml Erlenmeyer flasks or 1×55 and 1×39 250 ml, 1×32 and 1×27 500 ml, 20×1000 ml, 10×2000 ml or 1×5 5000 ml Schott Duran flasks.
Model 5075EL-PV-G-WS has a load capacity of 1×50 and 1×18 250 ml, 1×18, and 1×18 500 ml, 15×1000 ml, 7×2000 ml, 4×3000 ml or 3×5000 ml Erlenmeyer flasks or 1×55 and 1×39 250 ml, 1×32 and 1×27 500 ml, 20×1000 ml, 10×2000 ml or 1×5 5000 ml Schott Duran flasks.