OdorEroder® Absorbent Beads, SPEX CertiPrep
Supplier: Avantor
Certificates
About this item
OdorEroder® effectively neutralizes a wide range of offensive chemical odors and fumes in the lab, everything from aldehydes to xylenes, to cannabis.
Effective the in following areas:
- Hoods
- Waste Disposal Areas
- Lab Benches
- Chemical Storage Cabinets
- Glove Boxes
- Lab Refrigerators
- Other Odor-Causing Areas within a Lab
These non-toxic, environmentally safe purple beads are placed where odor causing chemicals in the air pass near the beads. When this occurs, the purple beads absorb and chemically transform the odor-causing chemicals into harmless compounds that remain trapped within the beads. As the purple beads absorb, they start turning brown. When a majority of the beads have turned brown, it is time to replace the OdorEroder®. Depending on the exposure, the OdorEroder® lasts up to three months.