Order Entry
Puerto Rico
ContactUsLinkComponent
RNAdvance Viral RNA Extraction Kit, Beckman Coulter
RNAdvance Viral RNA Extraction Kit, Beckman Coulter
Catalog # 76446-908
Supplier:  Beckman Coulter
CAS Number:  
undefined
RNAdvance Viral RNA Extraction Kit, Beckman Coulter
Catalog # 76446-908
Supplier:  Beckman Coulter
Supplier Number:  C63510
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    For research use, RNA isolation, RNA extraction, nucleic acid sample preparation, PCR
  • Size:
    768 preparations
  • Physical Form:
    Liquid
  • Cat. No.:
    76446-908

Specifications

About this item

Extract high-quality RNA from saliva, nasopharyngeal and oropharyngeal swab collection devices.

  • Produces viral RNA of high integrity and purity for use in PCR-based assays and NGS
  • Performance Limit of Detection (LoD) demonstrated at 1 copy/µL for viral RNA
  • Compatible with multiple sample inputs, including saliva and swab transport media
  • Automation-ready chemistry adaptable to high-throughput workflows
  • Uses saliva and swab in universal transport media as the starting sample material

Powered by Solid Phase Reversible Immobilization (SPRI) technology, the RNAdvance Viral kit uses paramagnetic beads to selectively bind nucleic acids for high yield and efficient removal of impurities. The scalable protocol can be performed in single tubes or 96-well plates with the flexibility to automate on a variety of liquid handling platforms.