ContactUsLinkComponent
Accessories for B.O.D Disposable Bottles, Environmental Express
Accessories for B.O.D Disposable Bottles, Environmental Express
Catalog # 76436-870
undefined
Accessories for B.O.D Disposable Bottles, Environmental Express
Catalog # 76436-870
Supplier Number:  D1050
Specifications
  • Description:
    Overcaps for B.O.D bottles, PE
  • Cat. No.:
    76436-870
Specifications
About this item
These overcaps are made of PE and fit snugly against the rim and shoulder of the disposable BOD bottles.
  • Ideal for 300 ml BOD bottles
The use of a cap over the mouth of a the BOD bottle to prevent evaporation is recommended by Standard Methods, and these overcaps do not fit the glass BOD bottles.