ContactUsLinkComponent
Transporter® Transport Swab, Amies Gel without Charcoal, Single Swab
Transporter® Transport Swab, Amies Gel without Charcoal, Single Swab
Catalog # 76441-348
Supplier:  Hardy Diagnostics
CAS Number:  
undefined
Transporter® Transport Swab, Amies Gel without Charcoal, Single Swab
Catalog # 76441-348
Supplier:  Hardy Diagnostics
Supplier Number:  4108BX
CAS Number:  
Specifications
  • Description:
    Amies gel without charcoal, plastic shaft, regular rayon tip, blue cap
  • Size:
    50/pack
  • Cat. No.:
    76441-348
Specifications
About this item
Transporter® Amies gel, without charcoal, single, plastic shaft, rayon tip swab and blue cap.
  • Available in two formats: liquid amies for aerobic culture amies agar gel (with and without charcoal) for aerobic and anaerobic culture
  • Unique tube shape preserves gel media
  • Maximum gel for anaerobe preservation
  • Available with or without charcoal
Amies media is Stuart's original media formula modified by replacing the glycerophosphate in the original formula for an inorganic phosphate buffer. The metabolism of glycerophosphate by coliform organisms and other gram negative rods, in Stuart's original formulation, resulted in proliferation of these organisms from throat, wound and fecal specimens.
Amies also found that NaCl at 0.3% w/v was optimal for the preservation of Neisseria gonorrhoeae. Calcium and magnesium salts were added in the belief that these ions were of importance in controlling the permeability of the bacterial cells and contributed to their survival.
Packaging: Pack of 50.