Order Entry
Puerto Rico
ContactUsLinkComponent
 
Phenol Red Broth with Sucrose, Hardy Diagnostics
Catalog # 76441-344
Supplier:  Hardy Diagnostics
CAS Number:  
undefined
Phenol Red Broth with Sucrose, Hardy Diagnostics
Catalog # 76441-344
Supplier:  Hardy Diagnostics
Supplier Number:  Y316
CAS Number:  

Specifications

  • Pack type:
    Tube with Durham tube
  • Description:
    Prepared media (Phenol red broth with sucrose)
  • Size:
    20/pack
  • Cat. No.:
    76441-344
  • Tube size:
    16×125 mm
  • Volume:
    10 ml

Specifications

About this item

For the differentiation of microorganisms by carbohydrate fermentation. Contains a durham tube for gas detection.

This phenol red carbohydrate test media is intended to be used in the differentiation of microorganisms based on fermentation reactions.

Packaging: Pack of 20.