Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Polysorbate 20, Multi-Compendial

Polysorbate 20, Multi-Compendial

Glass bottle, 200 ml
Catalog # JT4115-04
Supplier #:  4115-04
Polysorbate 20, Multi-Compendial
Catalog # JT4115-04
CAS Number:  9005-64-5
Polysorbate 20, Multi-Compendial
Polysorbate 20, Multi-Compendial

Glass bottle, 200 ml

CAS Number:  9005-64-5
Price
$437.30
Log in for availability
Each (200ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Density:1.11 g/cm³
  •  :Management of Change (MOC) category = R
  • Storage Temperature:Ambient
  • EINECS:500-018-3
Specifications

Specification Test Results

  • NF - Acid Value:
    ≤ 2.0
  • NF - Residue on Ignition:
    ≤ 0.25 %
  • NF - Dioxane:
    ≤ 10 ppm
  • NF - Ethylene Oxide:
    ≤ 1 ppm
  • NF - Hydroxyl Value, mg KOH/g:
    96 - 108
  • NF - Identification A:
    Passes Test
  • NF - Identification B:
    Passes Test
  • NF - Saponification Value, mg KOH/g:
    40 - 50
  • NF - Caproic Acid (Composition of Fatty Acids):
    ≤ 1.0 %
  • NF - Caprylic Acid (Composition of Fatty Acids):
    ≤ 10.0 %
  • NF - Capric Acid (Composition of Fatty Acids):
    ≤ 10.0 %
  • NF - Lauric Acid (Composition of Fatty Acids):
    40.0 - 60.0 %
  • NF - Myristic Acid (Composition of Fatty Acids):
    14.0 - 25.0 %
  • NF - Palmitic Acid (Composition of Fatty Acids):
    7.0 - 15.0 %
  • NF - Stearic Acid (Composition of Fatty Acids):
    ≤ 11.0 %
  • NF - Oleic Acid (Composition of Fatty Acids):
    ≤ 11.0 %
  • NF - Linoleic Acid (Composition of Fatty Acids):
    ≤ 3.0 %
  • NF - Peroxide Value:
    ≤ 5.0
  • NF - Water (H₂O):
    ≤ 3.0 %
  • EP/BP - Acid Value:
    ≤ 2.0
  • EP/BP - Total Ash:
    ≤ 0.25 %
  • EP/BP - Composition of Fatty Acids - Caproic Acid:
    ≤ 1.0 %
  • EP/BP - Composition of Fatty Acids - Caprylic Acid:
    ≤ 10.0 %
  • EP/BP - Composition of Fatty Acids - Capric Acid:
    ≤ 10.0 %
  • EP/BP - Composition of Fatty Acids - Lauric Acid:
    40.0 - 60.0 %
  • EP/BP - Composition of Fatty Acids - Myristic Acid:
    14.0 - 25.0 %
  • EP/BP -Composition of Fatty Acids - Palmitic Acid:
    7.0 - 15.0 %
  • EP/BP - Composition of Fatty Acids - Stearic Acid:
    ≤ 7.0 %
  • EP/BP - Composition of Fatty Acids - Oleic Acid:
    ≤ 11.0 %
  • EP/BP - Composition of Fatty Acids - Linoleic Acid:
    ≤ 3.0 %
  • EP - Identification A:
    Passes Test
  • EP - Identification D:
    Passes Test
  • EP/BP - Hydroxyl Value:
    96 - 108
  • EP - Peroxide Value:
    ≤ 10.0
  • EP - Saponification Value:
    40 - 50
  • EP - Water (H₂O):
    ≤ 3.0 %
  • EP - Ethylene Oxide:
    ≤ 1 ppm
  • EP - Dioxan:
    ≤ 10 ppm
  • Microbiological - Total Aerobic Microbial Count (cfu/g):
    ≤ 100
  • Microbiological - Escherichia Coli:
    Passes Test
  • Microbiological - Pseudomonas aeruginosa:
    Passes Test
  • Microbiological - Salmonella:
    Passes Test
  • Microbiological - Staphylococcus aureus:
    Passes Test
  • Microbiological - Total Yeast and Mold Count (cfu/g):
    ≤ 50
  • JPE - Identification 1:
    Passes Test
  • JPE - Identification 2:
    Passes Test
  • JPE - Identification 3:
    Passes Test
  • JPE - Moisture Content:
    ≤ 3.0 %
  • JPE - Acid Value:
    ≤ 4.0
  • JPE - Saponification Value (mg KOH/g):
    43 - 55
  • JPE - pH:
    4.0 - 7.0
  • JPE - Residue on Ignition:
    ≤ 0.25 %
  • JPE - Specific Gravity at 20°C:
    1.090 - 1.130
  • JPE - Viscosity (mm²/S):
    350 - 550
  • JPE - Heavy Metals (as Pb):
    ≤ 20 ppm
  • JPE - Arsenic (As):
    ≤ 2 ppm
  • ChP - Relative Density @ 20 C:
    1.09 - 1.12
  • ChP - Viscosity, mm2/s @ 25 C:
    250 - 400
  • ChP - Acid Value,meq/g:
    ≤ 2.0
  • ChP - Saponification Value:
    40 - 50
  • ChP - Hydroxyl Value:
    96 - 108
  • ChP - Peroxide Value:
    ≤ 10
  • ChP - Identification 1:
    Passes Test
  • ChP - Identification 2:
    Passes Test
  • ChP - Identification 3:
    Passes Test
  • ChP - Acidity or Alkalinity (pH):
    4.0 - 7.5
  • ChP - Color:
    Passes Test
  • ChP - Ethylene glycol:
    ≤ 0.01 %
  • ChP - Diethylene glycol:
    ≤ 0.01 %
  • ChP - Ethylene Oxide:
    ≤ 0.0001 %
  • ChP - Dioxane:
    ≤ 0.001 %
  • ChP - Water:
    ≤ 3.0 %
  • ChP - Heavy Metals:
    ≤ 10 ppm
  • ChP - Arsenic:
    ≤ 0.0002 %
  • ChP - Fatty Acid Composition: Lauric acid:
    40.0 - 60.0 %
  • ChP - Fatty Acid Composition: Myristic acid:
    14.0 - 25.0 %
  • ChP - Fatty Acid Composition: Palmitic acid:
    7.0 - 15.0 %
  • ChP - Fatty Acid Composition: Caproic acid:
    ≤ 1.0 %
  • ChP - Fatty Acid Composition: Caprylic acid:
    ≤ 10.0 %
  • ChP - Fatty Acid Composition: Capric acid:
    ≤ 10.0 %
  • ChP - Fatty Acid Composition: Stearic acid:
    ≤ 7.0 %
  • ChP - Fatty Acid Composition: Oleic acid:
    ≤ 11.0 %
  • ChP - Fatty Acid Composition: Linoleic acid:
    ≤ 3.0 %
  • Endotoxin Concentration (EU/mL):
    ≤ 10
  • Peroxide Value, meqO2/kg:
    ≤ 2.0
  • Appearance:
    Passes Test
  • pH of 5% solution at 25°C:
    5.0 - 7.0
  • Arsenic:
    ≤ 1 ppm
  • Ethylene glycol:
    ≤ 0.10 %
  • Diethylene glycol :
    ≤ 0.10 %

Specifications

  • Pack type:
    Glass bottle
  • Size:
    200 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.