
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99.0%, Biotech Reagent, J.T.Baker®
Catalog # JT4126-07
Supplier: AVANTOR PERFORMANCE MATERIALS US
CAS Number:
MOC: RegulatedCertificates
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:PE pail, screw top
- Size:12 kg
- Cat. No.:JT4126-07
Specifications
Specification Test Results
- Assay (NH₂C(CH₂OH)₃·HCl):99.0 - 101.0 %
- Appearance:Passes Test
- Absorbance of a 1M Solution - 260 nm:≤0.06
- Absorbance of a 1M Solution - 280 nm:≤0.06
- Absorbance of a 1M Solution - 400 nm:≤0.01
- Endotoxin Concentration (EU/g):≤2.5
- Identification (by IR):Passes Test
- Melting Point:147 - 153 °C
- pH of 0.5M Solution at 25°C:3.5 - 5.0
- Loss on Drying at 105°C:≤0.5 %
- Trace Impurities - Arsenic (As):≤10 ppm
- Trace Impurities - Calcium (Ca):≤10.0 ppm
- Trace Impurities - Copper (Cu):≤10 ppm
- Trace Impurities - Iron (Fe):≤10 ppm
- Trace Impurities - Lead (Pb):≤10 ppm
- Trace Impurities - Magnesium (Mg):≤10 ppm
Specification Test Results
About this item
:
:157.6 g/mol
:Management of Change (MOC) category = R
:GHS07
CAS Number:1185-53-1
Formula:NH₂C(CH₂OH)₃·HCl
MDL Number:MFCD00012590