ContactUsLinkComponent
Monarch® RNA Cleanup Wash Buffer, New England Biolabs
Monarch® RNA Cleanup Wash Buffer, New England Biolabs
Catalog # 76299-640
CAS Number:  
undefined
Monarch® RNA Cleanup Wash Buffer, New England Biolabs
Catalog # 76299-640
Supplier Number:  T2042L
CAS Number:  
Specifications
  • Description:
    Monarch® RNA cleanup wash buffer
  • Size:
    40 ml
  • Cat. No.:
    76299-640
Specifications
About this item
This is a component of the Monarch® RNA cleanup kits, is used to wash RNA after binding to ensure the removal of contaminants such as enzyme, NTPs, and short RNA fragments. It is supplied as a 5X concentrate and requires dilution with ethanol (≥95%) before use.
Monarch® kit components are available separately.
Delivery: The product is supplied in 2 bottles, each with 20 ml of concentrated buffer.