Cellvation DMSO Free Cryopreservation Solution, Protide Pharmaceuticals
Catalog # 76443-298
Supplier: PROTIDE PHARMACEUTICALS
CAS Number:
Specifications
- Shelf Life:18 months from DOM
- Endotoxin:<0.5 EU/ml at 1X
- Storage Temperature:2...8 °C
- Cat. No.:76443-298
- Volume:60 ml
Specifications
About this item
A unique cryopreservation solution that does not contain DMSO or fetal bovine serum. Cellvation™ is convenient to use and provides optimal recovery. Cells can be exposed to Cellvation™ for longer periods of time before the freezing process is initiated. Cellvation™ is ready to use and can be used for cells cultured in serum-free medium or with serum containing medium.
- Does not contain DMSO
- Sterile/Ready to use
Certifications: cGMP manufactured.