PCB Screening Standard for US EPA Method 8270, SPEX CertiPrep
SXDCBP-XH
:
- Concentration:5000 µg/ml
- Matrix:Toluene
- Pack type:Ampoule, amber
- CAS No.:2051-24-3
- Description:Decachlorobiphenyl
- Size:1 ml
- Description:Decachlorobiphenyl
- Cat. No.:SXDCBP-XH
Semivolatiles
US EPA Method 8270 is a general analytical method for the monitoring of semivolatiles in environmental and waste samples by GC/MS.