Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Chloroform Single-Component Organic Standard, SPEX CertiPrep

Chloroform Single-Component Organic Standard, SPEX CertiPrep

Chloroform, 1000 µg/ml, 1 ml, Methanol (P&T), Ampoule, amber
Catalog # SXS-865
Supplier:   SPEX CERTIPREP LLC
Supplier #:  S-865
Chloroform Single-Component Organic Standard, SPEX CertiPrep
Catalog # SXS-865
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Chloroform Single-Component Organic Standard, SPEX CertiPrep

Chloroform, 1000 µg/ml, 1 ml, Methanol (P&T), Ampoule, amber

CAS Number:  
Price
$55.11
Log in for availability
Each (1ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Single-Component organic standards.

Single-component standards are available as stock items. They are supplied in methanol, or in another solvent, where appropriate. Additional solvents include methylene chloride, acetone, benzene, hexane, acetonitrile, THF, and water. Please inquire about the matrix for a particular item when ordering, if the matrix is important to your application, a substitution may be possible.

Concentrations other than these, as well as custom matrices, are readily available, but a minimum order of five ampules may be required in some cases.

Specifications

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    Methanol (P&T)
  • Pack type:
    Ampoule, amber
  • Description:
    Chloroform
  • Size:
    1 ml
  • Cat. No.:
    SXS-865
  • Description:
    Chloroform
  • CAS No.:
    67-66-3

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.