Order Entry
Puerto Rico
ContactUsLinkComponent
Perylene Single-Component Organic Standard, SPEX CertiPrep
Perylene Single-Component Organic Standard, SPEX CertiPrep
Catalog # SXS-3005
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
Perylene Single-Component Organic Standard, SPEX CertiPrep
Catalog # SXS-3005
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  S-3005
CAS Number:  

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    Methylene chloride
  • Pack type:
    Ampoule, amber
  • CAS No.:
    198-55-0
  • Description:
    Perylene
  • Size:
    1 ml
  • Description:
    Perylene
  • Cat. No.:
    SXS-3005

Specifications

About this item

Single-Component organic standards.

Single-component standards are available as stock items. They are supplied in methanol, or in another solvent, where appropriate. Additional solvents include methylene chloride, acetone, benzene, hexane, acetonitrile, THF, and water. Please inquire about the matrix for a particular item when ordering, if the matrix is important to your application, a substitution may be possible.

Concentrations other than these, as well as custom matrices, are readily available, but a minimum order of five ampules may be required in some cases.