Order Entry
Puerto Rico
ContactUsLinkComponent
1,2-Dimethoxyethane Single-Component Organic Standard, SPEX CertiPrep
1,2-Dimethoxyethane Single-Component Organic Standard, SPEX CertiPrep
  SXS-1572
 :  
1,2-Dimethoxyethane Single-Component Organic Standard, SPEX CertiPrep
  SXS-1572
 :  S-1572
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Concentration:
    1000 µg/ml
  • Matrix:
    Methanol (P&T)
  • Pack type:
    Ampoule, amber
  • CAS No.:
    110-71-4
  • Description:
    1,2-Dimethoxyethane
  • Size:
    1 ml
  • Description:
    1,2-Dimethoxyethane
  • Cat. No.:
    SXS-1572

 

 

Single-Component organic standards.

Single-component standards are available as stock items. They are supplied in methanol, or in another solvent, where appropriate. Additional solvents include methylene chloride, acetone, benzene, hexane, acetonitrile, THF, and water. Please inquire about the matrix for a particular item when ordering, if the matrix is important to your application, a substitution may be possible.

Concentrations other than these, as well as custom matrices, are readily available, but a minimum order of five ampules may be required in some cases.