Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Captan Single-Component Organic Standard, SPEX CertiPrep

Captan Single-Component Organic Standard, SPEX CertiPrep

Captan Single-Component Organic Standard, SPEX CertiPrep
Supplier:  SPEX CERTIPREP LLC
Certificates
Captan Single-Component Organic Standard, SPEX CertiPrep

1 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Single-Component organic standards.

Single-component standards are available as stock items. They are supplied in methanol, or in another solvent, where appropriate. Additional solvents include methylene chloride, acetone, benzene, hexane, acetonitrile, THF, and water. Please inquire about the matrix for a particular item when ordering, if the matrix is important to your application, a substitution may be possible.

Concentrations other than these, as well as custom matrices, are readily available, but a minimum order of five ampules may be required in some cases.

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • SXS-720
  • SXS-720-ACN
  • Description:
  • Captan
  • Captan
  • CAS No.:
  • 133-06-2
  • 133-06-2

Specifications

Product Family Options

Product Information

  • Concentration
    Size
    Matrix
    Pack type
    Availability
    Price
  • SXS-720
    Captan
    Captan
    1000 µg/ml
    1 ml
    Acetone
    Ampoule, amber
    Discontinued
    Supplier Number: -
    Specifications:
    More
    Specifications
    Cat. No.
    SXS-720
    Description
    Captan
    CAS No.
    133-06-2
  • Captan
    1000 µg/ml
    1 ml
    Acetonitrile
    Glass bottle, amber
    Log in for availability
    Log in for availability
    Each (1ml)
    Each (1ml) - $133.27
    $133.27
    Each (1ml)
       $133.27

    Supplier Number: S-720-ACN
    Specifications:
    More
    Specifications
    Cat. No.
    SXS-720-ACN
    Description
    Captan
    CAS No.
    133-06-2

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.