Order Entry
Puerto Rico
ContactUsLinkComponent
Orthoboric acid 20 g/L, SimpleDist Snip & Pour™ Reagent
Orthoboric acid 20 g/L, SimpleDist Snip & Pour™ Reagent
Catalog # 76436-854
CAS Number:  
undefined
Orthoboric acid 20 g/L, SimpleDist Snip & Pour™ Reagent
Catalog # 76436-854
Supplier Number:  C7125
CAS Number:  

Specifications

  • Pack type:
    PTFE tube
  • Size:
    50×5 ml
  • Cat. No.:
    76436-854

Specifications

About this item

 :
 :61.83 g/mol
CAS Number:10043-35-3
Formula:B(OH)₃
MDL Number:MFCD00011337