Order Entry
Puerto Rico
ContactUsLinkComponent
Sodium hydroxide concentrate 1.25 N, Snip & Pour™
Sodium hydroxide concentrate 1.25 N, Snip & Pour™
Catalog # 76436-848
CAS Number:  
undefined
Sodium hydroxide concentrate 1.25 N, Snip & Pour™
Catalog # 76436-848
Supplier Number:  C7110
CAS Number:  

Specifications

  • Pack type:
    PFA tube
  • Size:
    50×10 ml
  • Cat. No.:
    76436-848

Specifications

About this item

 :
 :40 g/mol
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548