ContactUsLinkComponent
Primerdesign Ltd COVID-19 genesig Real-Time PCR assay
Catalog # 76436-783
Supplier:  PRIMER DESIGN LTD
CAS Number:  
Primerdesign Ltd COVID-19 genesig Real-Time PCR assay
Catalog # 76436-783
Supplier:  PRIMER DESIGN LTD
Supplier Number:  Z-PATH-COVID19
CAS Number:  
Specifications
  • Description:
    COVID19 FDA EUA Test Kit (US only)
  • Cat. No.:
    76436-783
Specifications
About this item
The genesig Real-Time PCR COVID-19 is authorised by the US Food and Drug Administration (FDA). 
  • Rapid detection and exclusive to the COVID-19 strain
  • Does not detect other related coronavirus strains
  • High priming efficiency
  • Accurate controls to confirm extraction, and assay validity
  • Lyophilised components for ambient shipping
  • Highly specific detection profile
Kit includes: oasig Lyophilised qPCR Master Mix, Primer and Probe Mix, Positive Control, Internal Extraction Control, No Template Control, Resuspension Buffers.