
Lithium tetraborate : lithium metaborate (67% : 33%) Pure Grade, fusion flux for XRF (X-ray fluorescence), for ICP and AAS
Catalog # SXFFB-6700-02
Supplier: Avantor
CAS Number:
Specifications
- Pack type:LDPE bottle
- Size:1 kg
- Cat. No.:SXFFB-6700-02
Specifications
About this item
:
:GHS07
MDL Number:MFCD01074946