ContactUsLinkComponent
Lithium tetraborate : lithium metaborate : lithium bromide (49.75% : 49.75% : 0.50%), fusion flux, ultrapure grade for XRF (X-ray fluorescence), for ICP and AAS
Irritant
Lithium tetraborate : lithium metaborate : lithium bromide (49.75% : 49.75% : 0.50%), fusion flux, ultrapure grade for XRF (X-ray fluorescence), for ICP and AAS
Catalog # SXFFB-5005-03
Supplier:  Avantor
CAS Number:  
Irritant
Lithium tetraborate : lithium metaborate : lithium bromide (49.75% : 49.75% : 0.50%), fusion flux, ultrapure grade for XRF (X-ray fluorescence), for ICP and AAS
Catalog # SXFFB-5005-03
Supplier:  Avantor
CAS Number:  
Specifications
  • Pack type:
    LDPE bottle
  • Size:
    1 kg
  • Cat. No.:
    SXFFB-5005-03
Specifications
About this item
 :
 :GHS07